
1-Stearoyl-rac-glycerol

Synonym(s):
1-Monooctadecanoyl-rac-glycerol, rac-Glycerol 1-stearate, DL-α-Stearin, Monostearin
Linear Formula:
CH3(CH2)16COOCH2CH(OH)CH2OH
CAS Number:
Molecular Weight:
358.56
Products
Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing