Skip to Content
Merck
All Photos(1)

Key Documents

700127P

Avanti

7-keto-25-hydroxycholesterol

3β,25-dihydroxy-5-cholesten-7-one, powder

Synonym(s):

cholest-5-ene-7-one-3β,25-diol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H44O3
CAS Number:
Molecular Weight:
416.64
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (700127P-1mg)
pkg of 1 × 5 mg (700127P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 700127P

shipped in

dry ice

storage temp.

−20°C

General description

7-keto-25-hydroxycholesterol synthesis from 7-ketocholesterol is catalyzed by the enzyme cholesterol 25-hydroxylase. 7β,25-dihydroxycholesterol (7β25OHC) is produced from the stereospecific reduction of 7k25OHC by the action of enzyme 11β-hydroxysteroid dehydrogenase type 1 (11β-HSD1). 11β-HSD type 2 enzyme catalyzes the reverse conversion of 7β25OHC to 7k25OHC.

Application

7-keto-25-hydroxycholesterol has been used as a substrate for 11β-hydroxysteroid dehydrogenase type 1 (11β-HSD1) expressed in human embryonic kidney (HEK)-293 cells to test the oxoreduction functionality of the enzyme. It has also been used to test its effect on Epstein-Barr virus-induced seven-transmembrane receptor (EB12) activation.

Biochem/physiol Actions

7-keto-25-hydroxycholesterol (7k25OHC) might interact with the cysteine-rich domain (CRD) of the smoothened (Smo) in the hedgehog pathway. However, 7-keto-25-OHC is incapable of activating Smo.

Packaging

5 mL Amber Glass Screw Cap Vial (700127P-1mg)
5 mL Amber Glass Screw Cap Vial (700127P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pengxiang Huang et al.
Cell, 166(5), 1176-1187 (2016-08-23)
In vertebrates, sterols are necessary for Hedgehog signaling, a pathway critical in embryogenesis and cancer. Sterols activate the membrane protein Smoothened by binding its extracellular, cysteine-rich domain (CRD). Major unanswered questions concern the nature of the endogenous, activating sterol and the
Benjamin R Myers et al.
Developmental cell, 26(4), 346-357 (2013-08-21)
Hedgehog (Hh) signaling during development and in postembryonic tissues requires activation of the 7TM oncoprotein Smoothened (Smo) by mechanisms that may involve endogenous lipidic modulators. Exogenous Smo ligands previously identified include the plant sterol cyclopamine (and its therapeutically useful synthetic
Katharina R Beck et al.
The Journal of steroid biochemistry and molecular biology, 190, 19-28 (2019-03-25)
Oxysterols are cholesterol metabolites derived through either autoxidation or enzymatic processes. They consist of a large family of bioactive lipids that have been associated with the progression of multiple pathologies. In order to unravel (patho-)physiological mechanisms involving oxysterols, it is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service