Merck
  • Annexin A2 is an independent prognostic biomarker for evaluating the malignant progression of laryngeal cancer.
Annexin A2 is an independent prognostic biomarker for evaluating the malignant progression of laryngeal cancer.
Experimental and therapeutic medicine (2017-12-30)
Shi Luo, Chubo Xie, Ping Wu, Jian He, Yaoyun Tang, Jing Xu, Suping Zhao
ABSTRACT
Due to the lack of a definite diagnosis, a frequent recurrence rate and resistance to chemotherapy or radiotherapy, the clinical outcome for patients with advanced laryngeal cancer has not improved over the last decade. Annexin A2 is associated with the invasion and metastasis of cancer cells. In the present study, it was demonstrated using differential proteomics analysis that Annexin A2 is highly expressed in laryngeal carcinoma tissues and this was confirmed using immunohistochemistry, which demonstrated that the expression of Annexin A2 in laryngeal carcinoma tissues was significantly higher than in healthy adjacent tissue. In addition, its potential predictive value in the prognosis of patients with laryngeal carcinoma was evaluated. The results demonstrated that Annexin A2 expression was significantly associated with tumor size, lymph node metastasis, distant metastasis and clinical stage. In addition, higher Annexin A2 expression was associated with a poor prognosis of patients with laryngeal cancer. Thus, the results of the present study indicate that Annexin A2 expression is an independent prognostic biomarker for evaluating the malignant progression of laryngeal cancer.
Sigma-Aldrich
Monoclonal Anti-ANXA2 antibody produced in mouse, clone 3E8-B6, purified immunoglobulin, buffered aqueous solution