Merck
  • Disinfection byproduct formation in drinking water sources: A case study of Yuqiao reservoir.
Disinfection byproduct formation in drinking water sources: A case study of Yuqiao reservoir.
Chemosphere (2017-04-27)
Hongyan Zhai, Xizhen He, Yan Zhang, Tingting Du, Adeyemi S Adeleye, Yao Li
ABSTRACT
This study investigated the potential formation of disinfection byproducts (DBPs) during chlorination and chloramination of 20 water samples collected from different points of Yuqiao reservoir in Tianjin, China. The concentrations of dissolved organic matter and ammonia decreased downstream the reservoir, while the specific UV absorbance (SUVA: the ratio of UV
Sigma-Aldrich
1,2-Dibromopropane, 97%