Merck
  • Satellite RNAs promote pancreatic oncogenic processes via the dysfunction of YBX1.
Satellite RNAs promote pancreatic oncogenic processes via the dysfunction of YBX1.
Nature communications (2016-09-27)
Takahiro Kishikawa, Motoyuki Otsuka, Takeshi Yoshikawa, Motoko Ohno, Hideaki Ijichi, Kazuhiko Koike
ABSTRACT
Highly repetitive tandem arrays at the centromeric and pericentromeric regions in chromosomes, previously considered silent, are actively transcribed, particularly in cancer. This aberrant expression occurs even in K-ras-mutated pancreatic intraepithelial neoplasia (PanIN) tissues, which are precancerous lesions. To examine the biological roles of the satellite RNAs in carcinogenesis, we construct mouse PanIN-derived cells expressing major satellite (MajSAT) RNA and show increased malignant properties. We find an increase in frequency of chromosomal instability and point mutations in both genomic and mitochondrial DNA. We identify Y-box binding protein 1 (YBX1) as a protein that binds to MajSAT RNA. MajSAT RNA inhibits the nuclear translocation of YBX1 under stress conditions, thus reducing its DNA-damage repair function. The forced expression of YBX1 significantly decreases the aberrant phenotypes. These findings indicate that during the early stage of cancer development, satellite transcripts may act as 'intrinsic mutagens' by inducing YBX1 dysfunction, which may be crucial in oncogenic processes.
Sigma-Aldrich
Anti-hnRNP-A2/B1 antibody, Mouse monoclonal, clone DP3B3, purified from hybridoma cell culture
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Anti-γ-Tubulin antibody, Mouse monoclonal, clone GTU-88, ascites fluid