Merck
3-methylglutaconic acidemia in Smith-Lemli-Opitz syndrome.
Pediatric research (1995-05-01)
R I Kelley, L Kratz
ABSTRACT
The branched-chain organic acid, 3-methylglutaconic acid, is an intermediate (as the CoA thioester) in the leucine degradative pathway as well as the mevalonate shunt, a pathway that links isoprenoid metabolism with mitochondrial acetyl-CoA metabolism. Because the majority of patients with abnormal 3-methylglutaconic aciduria or acidemia appear to have normal leucine metabolism, we have speculated that some patients with 3-methylglutaconic aciduria may have defects of polyisoprenoid or sterol biosynthesis leading to overflow of isoprenoid precursors to 3-methylglutaconate via the mevalonate shunt. We therefore measured plasma levels of 3-methylglutaconic acid in patients with a known defect of sterol biosynthesis, Smith-Lemli-Opitz syndrome, and found that the patients with the lowest cholesterol levels had abnormally increased plasma levels of 3-methylglutaconic acid, similar in magnitude to those of patients with idiopathic 3-methylglutaconic aciduria. This finding suggests that some patients with unexplained 3-methylglutaconic aciduria may have defects of isoprenoid or sterol biosynthesis underlying their abnormal organic aciduria.
Product Description
Supelco
3-Methylglutaconic acid, mixture of E and Z isomers, ≥98.0% (HPLC)