Merck
  • A cross-sectional study assessing dietary intake and physical activity in Canadian patients with nonalcoholic fatty liver disease vs healthy controls.
A cross-sectional study assessing dietary intake and physical activity in Canadian patients with nonalcoholic fatty liver disease vs healthy controls.
Journal of the Academy of Nutrition and Dietetics (2014-03-19)
Hannah E Da Silva, Bianca M Arendt, Seham A Noureldin, George Therapondos, Maha Guindi, Johane P Allard
ABSTRACT
Poor diet and a sedentary lifestyle can contribute to nonalcoholic fatty liver disease (NAFLD). Our aim was to compare diet and physical activity of patients with NAFLD and healthy controls with current recommendations. This was a cross-sectional study. Seventy-four patients with biopsy-proven NAFLD (33 simple steatosis and 41 steatohepatitis [NASH]) and 27 healthy controls participated between 2003 and 2011. Food records and activity logs were completed for 7 days. Results were compared with Dietary Reference Intakes and Canadian Physical Activity Guidelines. Plasma vitamin C was measured to assess food record accuracy. Intake/activity for each participant was compared with the recommendations and proportion of subjects not meeting the requirements was calculated. Groups were compared by Kruskal-Wallis and Mann-Whitney U test or z-test with Bonferroni adjustment. More patients with NASH (58.5%) were obese compared with patients with simple steatosis (24.2%) and healthy controls (7.4%; P<0.01). Patients with NAFLD showed more insulin resistance than healthy controls. The reported energy intake was below estimated requirements in all groups (P≤0.001). The proportion of subjects from each group exceeding acceptable energy intake from fat was as follows: simple steatosis: 27.3%; NASH: 46.3%; healthy controls: 63.0% (simple steatosis vs health controls; P<0.05) and from saturated fat: simple steatosis: 42.4%; NASH: 70.7%; healthy controls: 63.0% (simple steatosis vs. NASH; P<0.05). In each group, >80% of subjects did not consume enough linoleic or linolenic acid, vitamin D, and vitamin E, and >60% exceeded the upper intake level for sodium. Only 53.1% of patients with simple steatosis and 53.8% of patients with NASH, but 84.6% of healthy controls, met recommendations for physical activity (P=0.020). Plasma vitamin C was normal, similar among groups, and correlated with vitamin C intakes. All participants followed a similar Western diet with high fat and sodium intakes and suboptimal micronutrient intakes. However, physical activity was lower in NAFLD compared with healthy controls and was associated with higher body mass index and insulin resistance.
Product Description
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
USP
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
(+)-Sodium L-ascorbate, SAJ special grade, ≥99.0%
Supelco
L-Ascorbic acid, analytical standard
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
(+)-Sodium L-ascorbate, crystalline, ≥98%
Sigma-Aldrich
(+)-Sodium L-ascorbate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
(+)-Sodium L-ascorbate, BioXtra, ≥99.0% (NT)
Supelco
Sodium ascorbate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sodium ascorbate, European Pharmacopoeia (EP) Reference Standard