Merck
  • Rapid induction of alternative lengthening of telomeres by depletion of the histone chaperone ASF1.
Rapid induction of alternative lengthening of telomeres by depletion of the histone chaperone ASF1.
Nature structural & molecular biology (2014-01-15)
Roderick J O'Sullivan, Nausica Arnoult, Daniel H Lackner, Liana Oganesian, Candy Haggblom, Armelle Corpet, Genevieve Almouzni, Jan Karlseder
ABSTRACT
The mechanism of activation of the alternative lengthening of telomeres (ALT) pathway of mammalian chromosome-end maintenance has been unclear. We have now discovered that co-depletion of the histone chaperones ASF1a and ASF1b in human cells induced all hallmarks of ALT in both primary and cancer cells. These included the formation of ALT-associated PML (promyelocytic leukemia) bodies (APBs), the presence of extrachromosomal telomeric DNA species, an elevated frequency of telomeric sister chromatid exchanges (t-SCE) events and intertelomeric exchange of an integrated tag. The induction of ALT characteristics in this setting led to the simultaneous suppression of telomerase. We determined that ALT induction is positively regulated by the proteins RAD17 and BLM and negatively regulated by EXO1 and DNA2. The induction of ALT phenotypes as a consequence of ASF1 depletion strongly supports the hypothesis that ALT is a consequence of histone management dysfunction.
Sigma-Aldrich
Anti-γ-Tubulin antibody, Mouse monoclonal, clone GTU-88, ascites fluid
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPrice
PBMR151872-100MG100 mg
Ships in 14 days - from Aldrich Partner.
€338.72
PBMR151872-250MG250 mg
Ships in 14 days - from Aldrich Partner.
€422.34
PBMR151872-500MG500 mg
Ships in 14 days - from Aldrich Partner.
€531.06
PBMR151872-1G1 g
Ships in 14 days - from Aldrich Partner.
€672.38
PBMR151872-2G2 g
Ships in 14 days - from Aldrich Partner.
€1,192.89
To order products, please contact your local dealer.
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPrice
PBMR151872-100MG100 mg
Ships in 14 days - from Aldrich Partner.
€338.72
PBMR151872-250MG250 mg
Ships in 14 days - from Aldrich Partner.
€422.34
PBMR151872-500MG500 mg
Ships in 14 days - from Aldrich Partner.
€531.06
PBMR151872-1G1 g
Ships in 14 days - from Aldrich Partner.
€672.38
PBMR151872-2G2 g
Ships in 14 days - from Aldrich Partner.
€1,192.89
To order products, please contact your local dealer.