Merck
  • Emergence of rifampicin, tigecycline, and colistin-resistant Acinetobacter baumannii in Iran; spreading of MDR strains of novel International Clone variants.
Emergence of rifampicin, tigecycline, and colistin-resistant Acinetobacter baumannii in Iran; spreading of MDR strains of novel International Clone variants.
Microbial drug resistance (Larchmont, N.Y.) (2013-06-19)
Abbas Bahador, Mohammad Taheri, Babak Pourakbari, Zahra Hashemizadeh, Hossein Rostami, Noormohamad Mansoori, Reza Raoofian
ABSTRACT
Multidrug-resistant Acinetobacter baumannii infections are serious challenges for clinicians because of A. baumannii propensity to acquire resistance to a wide spectrum of antimicrobial agents. In this study, 91 A. baumannii isolates from patients in tertiary intensive care units of three university hospitals in the north, central, and south of Iran were selected and tested for susceptibility to 22 antimicrobials; amplified restriction fragment polymorphism and multiplex polymerase chain reaction methods were used to determine genetic relationships and International Clone (IC) of A. baumannii isolates, respectively. Twenty-four genotypes were identified in A. baumannii isolates. About 91.2% of isolates categorized into 4 distinct clusters; one was more heterogeneous and observed across the three locations. A considerable number of the isolates (27.5%) belonged to the novel IC variant, sequence group 7 (SG7), which was geographically widespread in three locations. The drug resistance pattern showed that 14.2%, 20%, and 77% of the A. baumannii isolates were resistant to colistin, tigecycline, and rifampicin, respectively. Nine percent of isolates (8) showed simultaneous resistance to colistin, rifampicin, and tigecycline. Interestingly, all of them were susceptible to ampicillin-sulbactam and/or tobramycin. According to our results, SG7 could be considered as a pan-Iranian clone.
Product Description
Sulbactam sodium, European Pharmacopoeia (EP) Reference Standard
Sultamicillin, European Pharmacopoeia (EP) Reference Standard
Sultamicillin for peak identification, European Pharmacopoeia (EP) Reference Standard