Merck
  • Intramolecular direct dehydrohalide coupling promoted by KO(t)Bu: total synthesis of Amaryllidaceae alkaloids anhydrolycorinone and oxoassoanine.
Intramolecular direct dehydrohalide coupling promoted by KO(t)Bu: total synthesis of Amaryllidaceae alkaloids anhydrolycorinone and oxoassoanine.
Organic letters (2012-08-16)
Subhadip De, Santanu Ghosh, Subhajit Bhunia, Javeed Ahmad Sheikh, Alakesh Bisai
ABSTRACT
A transition-metal-free intramolecular dehydrohalide coupling via intramolecular homolytic aromatic substitution (HAS) with aryl radicals has been developed in the presence of potassium tert-butoxide and an organic molecule as the catalyst. The methodology has been applied to a concise synthesis of Amaryllidaceae alkaloids viz. oxoassoanine (1b), anhydrolycorinone (1d), and other related structures. Interestingly, the method also works only in the presence of potassium tert-butoxide.
Sigma-Aldrich
Potassium tert-butoxide solution, 1.0 M in THF
Sigma-Aldrich
Potassium tert-butoxide, sublimed grade, 99.99% trace metals basis
Sigma-Aldrich
Potassium tert-butoxide, reagent grade, ≥98%