Merck
Interdigital ulcer: an unusual presentation of Candida infection.
Mycoses (2011-05-28)
Di-Qing Luo, Wei Yang, Liang-Cai Wu, Jun-Hua Liu, Wen-Na Chen
ABSTRACT
Interdigital ulcer is an exceptionally rare condition while erosio interdigitalis blastomycetica is common for candidiasis. Four Chinese patients with Candida interdigital ulcers were reported. The exudates were examined directly and cultured for fungi. Skin biopsies were stained with haematoxylin-eosin and periodic acid Schiff. There were a man and three women (age range: 34-56 years) who presented with 1- to 3-month history of chronic cutaneous ulcer on the interdigital web of hand or foot. The lesions were located on hand for one woman, and on the left foot for the rest. The patients had poor response to the previous treatment of topical steroids and oral antimicrobials. Candida albicans was isolated from a man and two women, Candida tropicalis from another woman. Biopsy specimens revealed yeast and mycelium as well as inflammatory infiltrate in necrotic tissue in two patients; only inflammatory cells in the other two. The patients had complete remission with oral itraconazole and topical bifonazole cream therapy for 3- to 5-week. Candida species may cause interdigital ulcer on hand or foot. Oral itraconazole and topical bifonazole may be an optional therapy for such an ulcer.
Product Description
Bifonazole, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Bifonazole, ≥98% (HPLC)
Supelco
Bifonazole, Pharmaceutical Secondary Standard; Certified Reference Material