Merck
  • Resolution and sensitivity aspects of ultrafast J-resolved 2D NMR spectra.
Resolution and sensitivity aspects of ultrafast J-resolved 2D NMR spectra.
Journal of magnetic resonance (San Diego, Calif. : 1997) (2007-12-18)
Patrick Giraudeau, Serge Akoka
ABSTRACT
Recent ultrafast techniques enable nD NMR spectra to be obtained in a single scan. However, resolution enhancement in the ultrafast domain leads to important sensitivity losses and lineshape distortions. In order to understand better resolution and spatial encoding aspects of continuous phase-encoding schemes, a theoretical and experimental comparison of different excitation patterns is carried out. Molecular diffusion appears to be the main cause of signal-to-noise ratio decrease, and a multi-echo excitation scheme is proposed to limit its effects when a good resolution is needed. Results obtained on 2D J-resolved spectra are presented.
Sigma-Aldrich
3-Bromopropionic acid, 97%