Merck
  • Steric and electronic factors influencing recognition by a simple, charge neutral norbornene based anion receptor.
Steric and electronic factors influencing recognition by a simple, charge neutral norbornene based anion receptor.
Organic & biomolecular chemistry (2007-04-28)
Adam J Lowe, Gail A Dyson, Frederick M Pfeffer
ABSTRACT
Based on (1)H NMR studies, subtle electronic factors rather than pre-organisation dictate the binding stoichiometry of the new, norbornene based, anion hosts 1 and 2 with acetate, however, the binding of dihydrogenphosphate appears to be based solely on steric constraints.
Sigma-Aldrich
Dimethyl maleate, 96%