Merck
  • An efficient synthesis of 2,3-dideoxy-alpha,beta-unsaturated carbohydrate enals by mixed Lewis acid (HfCl4 and ZnI2) catalyzed hydration of glycals.
An efficient synthesis of 2,3-dideoxy-alpha,beta-unsaturated carbohydrate enals by mixed Lewis acid (HfCl4 and ZnI2) catalyzed hydration of glycals.
Carbohydrate research (2006-03-28)
Mohammad Saquib, Ram Sagar, Arun K Shaw
ABSTRACT
A new, efficient method has been developed for converting acyl-, arylalkyl- and alkyl-protected glycals into corresponding 2,3-dideoxy-alpha,beta-unsaturated carbohydrate enals utilizing the in situ generated push-pull effect resulting from the synergistic combination of HfCl4 and ZnI2 in catalytic amounts. This new procedure eliminates the use of highly toxic Hg2+ ions and acidic conditions (0.01-0.02 N H2SO4), besides radically shortening the reaction time.
Sigma-Aldrich
Zinc iodide, ≥99.99% trace metals basis
Sigma-Aldrich
Zinc iodide, purum p.a., ≥98.0% (AT)
Sigma-Aldrich
Zinc iodide, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Zinc iodide, ≥98%