Merck
  • Simultaneous analysis of clozapine, clomipramine and their metabolites by reversed-phase liquid chromatography.
Simultaneous analysis of clozapine, clomipramine and their metabolites by reversed-phase liquid chromatography.
Progress in neuro-psychopharmacology & biological psychiatry (2001-05-24)
L Palego, L Dell'Osso, D Marazziti, L Biondi, N Sarno, A Ciapparelli, A Giromella, G Giannaccini, LucacchiniA, G B Cassano
ABSTRACT
1. The authors present here a sensitive and rapid reversed-phase liquid chromatographic method which enables the simultaneous analysis in plasma of two different drugs and their metabolites: the atypical neuroleptic clozapine and the tricyclic antidepressant clomipramine. 2. Samples and the internal standard (dibenzepine) were extracted through automated solid-phase procedure, evaporated dryness and injected into the chromatograph. Mobile phase was a mixture of water and acetonitrile (63:37, v:v) containing TEMED and triethylamine. The total chromatographic time was of 14 min and analyte peaks were detected by means of an ultraviolet spectrophotometer preset at 254 nm. 3. Results revealed an assay sensitivity of 5 microg/L for clozapine or norclozapine and of 10 microg/L for clomipramine and desmethylclomipramine. Recoveries for these drugs and their metabolites were more than 60% and their coefficient of variation (within day and day-to-day) ranged from 1.3 % to 2.5 %. In spiked plasma, within day and day-to-day coefficients of variability (CV) were less than 5%. The simultaneous evaluation of these two drugs with adequate sensitivity and precision makes it particularly useful for therapeutic drug monitoring during mono- or polypharmacotherapy.
Product Description
Supelco
N-Desmethylclomipramine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Norclomipramine hydrochloride, ≥98% (TLC), powder