Merck
Mechanism of interdigitation formation at apical boundary of MDCK cell.
iScience (2023-05-30)
Shintaro Miyazaki, Tetsuhisa Otani, Kei Sugihara, Toshihiko Fujimori, Mikio Furuse, Takashi Miura
ABSTRACT
It has been reported that the MDCK cell tight junction shows stochastic fluctuation and forms the interdigitation structure, but the mechanism of the pattern formation remains to be elucidated. In the present study, we first quantified the shape of the cell-cell boundary at the initial phase of pattern formation. We found that the Fourier transform of the boundary shape shows linearity in the log-log plot, indicating the existence of scaling. Next, we tested several working hypotheses and found that the Edwards-Wilkinson equation, which consists of stochastic movement and boundary shortening, can reproduce the scaling property. Next, we examined the molecular nature of stochastic movement and found that myosin light chain puncta may be responsible. Quantification of boundary shortening indicates that mechanical property change may also play some role. Physiological meaning and scaling properties of the cell-cell boundary are discussed.
Sigma-Aldrich
CK-666, ≥98% (HPLC), powder
Sigma-Aldrich
Monoclonal Anti-Vinculin antibody produced in mouse, clone VIN-11-5, ascites fluid
Sigma-Aldrich
Formin FH2 Domain Inhibitor, SMIFH2, The Formin FH2 Domain Inhibitor, SMIFH2, also referenced under CAS 340316-62-3, controls the biological activity of Formin FH2 Domain. This small molecule/inhibitor is primarily used for Membrane applications.