Merck
Fluorescent labeling of s2T-incorporated DNA and m5s2U-modified RNA.
Nucleosides, nucleotides & nucleic acids (2021-06-29)
Ping Yu, Honglin Zhou, Yuanyuan Li, Zhifeng Du, Rui Wang
ABSTRACT
We report herein comprehensive investigations of alkylation/sulfur exchange reactions of sulfur-containing substrates including nucleosides such as s2U, m5s2U, s4U, s2A and s2T-incorporated DNA enable by comprehensive screenings of the reagents (2a-2h). It has been proven that iodoacetamide (2a) displays the most promising feasibility toward sulfur-containing substrates including s2T, s2U, m5s2U, s4U and s2A. In sharp contrast, the alkylation process with S-benzyl methanethiosulfonate (BMTS, 2h) displays the best application potential only for s4U. Based on these results, the fluorescent labeling of s2T-incorporated DNA and m5s2U-modified RNA has been achieved.Supplemental data for this article is available online at https://doi.org/10.1080/15257770.2021.1942044 .
Sigma-Aldrich
HMP-alkyne, ≥95%
Sigma-Aldrich
SuTEx1-alkyne, ≥95%
Sigma-Aldrich
OxMet2-alkyne
Sigma-Aldrich
CP-alkyne, ≥95%