Merck
  • NELL1 is a target antigen in malignancy-associated membranous nephropathy.
NELL1 is a target antigen in malignancy-associated membranous nephropathy.
Kidney international (2020-08-24)
Tiffany N Caza, Samar I Hassen, Zeljko Dvanajscak, Michael Kuperman, Rick Edmondson, Christian Herzog, Aaron Storey, John Arthur, L Nicholas Cossey, Shree G Sharma, Daniel J Kenan, Christopher P Larsen, Tiffany N Caza, Samar I Hassen, Zeljko Dvanajscak, Michael Kuperman, Rick Edmondson, Christian Herzog, Aaron Storey, John Arthur, L Nicholas Cossey, Shree G Sharma, Daniel J Kenan, Christopher P Larsen
ABSTRACT
Patients with membranous nephropathy have an increased risk of malignancy compared to the general population, but the target antigen for malignancy-associated membranous nephropathy is unknown. To explore this, we utilized mass spectrometry for antigen discovery in malignancy-associated membranous nephropathy examining immune complexes eluted from frozen kidney biopsy tissue using protein G bead immunoglobulin capture. Antigen discovery was performed comparing cases of membranous nephropathy of unknown and known type. Mass spectrophotometric analysis revealed that nerve epidermal growth factor-like 1 (NELL1) immune complexes were uniquely present within the biopsy tissue in membranous nephropathy. Additional NELL1-positive cases were subsequently identified by immunofluorescence. In a consecutive series, 3.8% of PLA2R- and THSD7A-negative cases were NELL1-positive. These NELL1-positive cases had segmental to incomplete IgG capillary loop staining (93.4%) and dominant or co-dominant IgG1-subclass staining (95.5%). The mean age of patients with NELL1-positive membranous nephropathy was 66.8 years, with a slight male predominance (58.2%) and 33% had concurrent malignancy. Compared with PLA2R- and THSD7A-positive cases of membranous nephropathy, there was a greater proportion of cases with malignancies in the NELL1-associated group. Thus, NELL1-associated membranous nephropathy has a unique histopathology characterized by incomplete capillary loop staining, IgG1-predominance, and is more often associated with malignancy than other known types of membranous nephropathy.
Sigma-Aldrich
Monoclonal Anti-THSD7A antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL3779, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-PLA2R1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution