Merck
  • Insulin-Driven PI3K-AKT Signaling in the Hepatocyte Is Mediated by Redundant PI3Kα and PI3Kβ Activities and Is Promoted by RAS.
Insulin-Driven PI3K-AKT Signaling in the Hepatocyte Is Mediated by Redundant PI3Kα and PI3Kβ Activities and Is Promoted by RAS.
Cell metabolism (2019-04-16)
Angela Molinaro, Barbara Becattini, Arianna Mazzoli, Augusto Bleve, Lucia Radici, Ingela Maxvall, Victoria Rotter Sopasakis, Antonio Molinaro, Fredrik Bäckhed, Giovanni Solinas
ABSTRACT
Phosphatidylinositol-3-kinase (PI3K) activity is aberrant in tumors, and PI3K inhibitors are investigated as cancer therapeutics. PI3K signaling mediates insulin action in metabolism, but the role of PI3K isoforms in insulin signaling remains unresolved. Defining the role of PI3K isoforms in insulin signaling is necessary for a mechanistic understanding of insulin action and to develop PI3K inhibitors with optimal therapeutic index. We show that insulin-driven PI3K-AKT signaling depends on redundant PI3Kα and PI3Kβ activities, whereas PI3Kδ and PI3Kγ are largely dispensable. We have also found that RAS activity promotes AKT phosphorylation in insulin-stimulated hepatocytes and that promotion of insulin-driven AKT phosphorylation by RAS depends on PI3Kα. These findings reveal the detailed mechanism by which insulin activates AKT, providing an improved mechanistic understanding of insulin signaling. This improved model for insulin signaling predicts that isoform-selective PI3K inhibitors discriminating between PI3Kα and PI3Kβ should be dosed below their hyperglycemic threshold to achieve isoform selectivity.
Sigma-Aldrich
Guanidine thiocyanate, ≥97% (titration)
Sigma-Aldrich
Anti-IRS1 Antibody, Upstate®, from rabbit
Sigma-Aldrich
Collagenase from Clostridium histolyticum, suitable for release of physiologically active rat hepatocytes, Type IV, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
Anti-HA antibody, Mouse monoclonal, clone HA-7, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Phosphotyrosine Antibody, clone 4G10®, clone 4G10®, Upstate®, from mouse