Merck
  • Exercise intervention alters HDL subclass distribution and function in obese women.
Exercise intervention alters HDL subclass distribution and function in obese women.
Lipids in health and disease (2018-10-12)
Nicholas J Woudberg, Amy E Mendham, Arieh A Katz, Julia H Goedecke, Sandrine Lecour
ABSTRACT
Obesity is associated with a change in high-density lipoprotein (HDL) function and subclass. Exercise training reduces cardiovascular risk in obese patients. We aimed to explore the effect of an exercise training stimulus on HDL functionality and subclass in obese women. Thirty-two obese black South African women were randomly assigned to exercise (combined aerobic and resistance exercise) or control (no exercise) conditions for 12-weeks. Pre- and post-testing included venous blood sampling for analysis of lipid profile and HDL functionality, by measuring cellular cholesterol efflux capacity, reduction in endothelial vascular cell adhesion molecule (VCAM) expression (anti-inflammatory function), paraoxonase (PON) (antioxidative function) and platelet activating factor acetylhydrolase (PAF-AH) activities (anti-thrombotic function). PON-1 and PAF-AH expression were determined in serum and in isolated HDL using Western blotting. Levels of large, intermediate and small HDL subclasses were measured using the Lipoprint® system. Exercise training resulted in a decrease in body mass index (- 1.0 ± 0.5% vs + 1.2 ± 0.6%, p = 0.010), PON activity (- 8.7 ± 2.4% vs + 1.1 ± 3.0%, p = 0.021), PAF-AH serum expression (- 22.1 ± 8.0% vs + 16.9 ± 9.8, p = 0.002), and the distribution of small HDL subclasses (- 10.1 ± 5.4% vs + 15.7 ± 6.6%, p = 0.004) compared to controls. Exercise did not alter HDL cellular cholesterol efflux capacity and anti-inflammatory function. These results demonstrate the potential for exercise training to modify HDL subclass distribution and HDL function in obese women. Clinical trials number: PACTR201711002789113 .
Sigma-Aldrich
Paraoxon-ethyl, ≥90%, oil
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture