Merck
  • New insights into the regulatory function of CYFIP1 in the context of WAVE- and FMRP-containing complexes.
New insights into the regulatory function of CYFIP1 in the context of WAVE- and FMRP-containing complexes.
Disease models & mechanisms (2017-02-12)
Sabiha Abekhoukh, H Bahar Sahin, Mauro Grossi, Samantha Zongaro, Thomas Maurin, Irene Madrigal, Daniele Kazue-Sugioka, Annick Raas-Rothschild, Mohamed Doulazmi, Pilar Carrera, Andrea Stachon, Steven Scherer, Maria Rita Drula Do Nascimento, Alain Trembleau, Ignacio Arroyo, Peter Szatmari, Isabel M Smith, Montserrat Milà, Adam C Smith, Angela Giangrande, Isabelle Caillé, Barbara Bardoni
ABSTRACT
Cytoplasmic FMRP interacting protein 1 (CYFIP1) is a candidate gene for intellectual disability (ID), autism, schizophrenia and epilepsy. It is a member of a family of proteins that is highly conserved during evolution, sharing high homology with its Drosophila homolog, dCYFIP. CYFIP1 interacts with the Fragile X mental retardation protein (FMRP, encoded by the FMR1 gene), whose absence causes Fragile X syndrome, and with the translation initiation factor eIF4E. It is a member of the WAVE regulatory complex (WRC), thus representing a link between translational regulation and the actin cytoskeleton. Here, we present data showing a correlation between mRNA levels of CYFIP1 and other members of the WRC. This suggests a tight regulation of the levels of the WRC members, not only by post-translational mechanisms, as previously hypothesized. Moreover, we studied the impact of loss of function of both CYFIP1 and FMRP on neuronal growth and differentiation in two animal models - fly and mouse. We show that these two proteins antagonize each other's function not only during neuromuscular junction growth in the fly but also during new neuronal differentiation in the olfactory bulb of adult mice. Mechanistically, FMRP and CYFIP1 modulate mTor signaling in an antagonistic manner, likely via independent pathways, supporting the results obtained in mouse as well as in fly at the morphological level. Collectively, our results illustrate a new model to explain the cellular roles of FMRP and CYFIP1 and the molecular significance of their interaction.
Sigma-Aldrich
Anti-PI3 Kinase Antibody, p110β, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid