
Vardenafil hydrochloride

Synonym(s):
Vardenafil hydrochloride trihydrate
Empirical Formula (Hill Notation):
C23H32N6O4S · HCl · 3H2O
CAS Number:
Molecular Weight:
579.11
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Special Grade
Showing 1-2 of 2