
Tween® 20 Solution

Synonym(s):
TWEEN® 20, Polyethylene glycol sorbitan monolaurate, Polyoxyethylenesorbitan monolaurate
CAS Number:
Products
Product Category
Brand
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing