Skip to Content
Merck
trans-Zeatin suitable for plant cell culture, BioReagent, ≥97%

trans-Zeatin

trans-Zeatin suitable for plant cell culture, BioReagent, ≥97%
Synonym(s):
6-(4-Hydroxy-3-methylbut-2-enylamino)purine, N6-(4-Hydroxy-3-methyl-2-buten-1-yl)adenine
Empirical Formula (Hill Notation):
C10H13N5O
CAS Number:
Molecular Weight:
219.24

Products

Brand
Formula Weight
Product Line
Purity
Application
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing