
Sodium Trimetaphosphate

Synonym(s):
Trisodium trimetaphosphate, Sodium cyclotriphosphate
Linear Formula:
Na3P3O9
CAS Number:
Molecular Weight:
305.89
Products
Brand
Formula Weight
Agency
Format
Quality Segment
Application
Package Size
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing