
Sesamin

Synonym(s):
Fagarol
Empirical Formula (Hill Notation):
C20H18O6
CAS Number:
Molecular Weight:
354.35
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing