Skip to Content
Merck
(R)-(−)-6-Methoxy-α-methyl-2-naphthaleneacetic acid 98%

(R)-(−)-6-Methoxy-α-methyl-2-naphthaleneacetic acid

(R)-(−)-6-Methoxy-α-methyl-2-naphthaleneacetic acid 98%
Synonym(s):
(2R)-2-(6-Methoxy-2-naphthyl)propionic acid, (2R)-2-(6-Methoxynaphthalen-2-yl)propanoic acid, (R)-Naproxen
Linear Formula:
CH3OC10H6CH(CH3)CO2H
CAS Number:
Molecular Weight:
230.26
EC Number:

Products

Product Category
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing