
Quinidine sulfate

Synonym(s):
Quinidine sulfate salt
Linear Formula:
C40H48N4O4 · H2SO4
CAS Number:
Molecular Weight:
746.91
Products
Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing