
Pyrex® gas-washing bottle, fritted cylinder, 29/42 ST stopper

Synonym(s):
Corning® 31770
Products
Product Category
Manufacturer
Capacity (mL)
Diameter (mm)
Cap
Bottle material
Color
Vial Bottle Material
Vial Bottle Color
Package Size
Pore size (µm)
Showing 1-5 of 5
Compare
Product Number
Description
Pricing