Potassium aluminium sulfate dodecahydrate
Synonym(s):
Aluminium potassium sulfate dodecahydrate
Linear Formula:
KAl(SO4)2 · 12 H2O
CAS Number:
Molecular Weight:
474.39
Products
Brand
Formula Weight
Product Line
Showing 1-1 of 1
Compare
Product Number
Description
Pricing