
PIPES disodium salt

Synonym(s):
1,4-Piperazinediethanesulfonic acid disodium salt, Piperazine-1,4-bis(2-ethanesulfonic acid) disodium salt, PIPES disodium salt
Empirical Formula (Hill Notation):
C8H16N2Na2O6S2
CAS Number:
Molecular Weight:
346.33
Products
Product Category
Brand
Biological Source
Feature
Formula Weight
pH Value
Physical Form
pK Value
Product Line
Purity
Quality Segment
Application
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing