
Phospho-L-Tyrosine Disodium Salt

Synonym(s):
Phospho-L-tyrosine disodium salt, PTyr, Phospho-Tyrosine, Phosphotyrosine
Empirical Formula (Hill Notation):
C9H10NNa2O6P
CAS Number:
Molecular Weight:
305.13
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Application
Showing 1-2 of 2