
Perfluoro-1-heptanesulfonic acid

Synonym(s):
1-Perfluoroheptanesulfonic acid, 1,1,2,2,3,3,4,4,5,5,6,6,7,7,7-Pentadecafluoro-1-heptanesulfonic acid
Empirical Formula (Hill Notation):
C7HF15O3S
CAS Number:
Molecular Weight:
450.12
Products
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing