Skip to Content
Merck
Pemetrexed disodium heptahydrate ≥98% (HPLC)

Pemetrexed disodium heptahydrate

Pemetrexed disodium heptahydrate ≥98% (HPLC)
Synonym(s):
HSDB 7316, LY 231,514, LY 231514, N-(4-(2-(2-Amino-3,4-dihydro-4-oxo-7H-pyrrolo(2,3-d)pyrimdin-5-yl)ethyl)benzoyl)glutamic acid sodium salt hydrate, N-(4-(2-(2-Amino-4,7-dihydro-4-oxo-1H-pyrrolo(2,3-d)pyrimidin-5-yl)ethyl)benzoyl)-L-glutamic acid sodium salt hydrate, UNII-04Q9AIZ7NO
Empirical Formula (Hill Notation):
C20H19N5Na2O6 · 7H2O
CAS Number:
Molecular Weight:
597.48

Products

Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Purity
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade