
Oxcarbazepine Related Compound E

Synonym(s):
5,11-Dihydro-10H-dibenz[b,f]azepin-10-one
Empirical Formula (Hill Notation):
C14H11NO
CAS Number:
Molecular Weight:
209.24
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2