
N-Nitrosomethylphenylamine (NMPA)

Synonym(s):
N-Methyl-N-nitrosoaniline, N-Methyl-N-nitrosobenzenamine, N-Methyl-N-phenylnitrosamine, NMPA
Empirical Formula (Hill Notation):
C7H8N2O
CAS Number:
Molecular Weight:
136.15
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Showing 1-2 of 2