
N-Nitrosodiisopropylamine (NDIPA)

Synonym(s):
N-Isopropyl-N-nitrosoisopropylamine, DIPNA, NDIP
Empirical Formula (Hill Notation):
C6H14N2O
CAS Number:
Molecular Weight:
130.19
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Showing 1-2 of 2