
N-Nitrosodibenzylamine

Synonym(s):
NDBzA
Empirical Formula (Hill Notation):
C14H14N2O
CAS Number:
Molecular Weight:
226.27
Products
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2