
Nitrotetrazolium Blue chloride

Synonym(s):
2,2′-bis(4-Nitrophenyl)-5,5′-diphenyl-3,3′-(3,3′-dimethoxy-4,4′-diphenylene)ditetrazolium chloride, 3,3′-(3,3′-Dimethoxy-4,4′-biphenylene)bis[2-(4-nitrophenyl)-5-phenyl-2H-tetrazolium chloride], p-Nitro-Blue tetrazolium chloride, p-Nitrotetrazolium blue, NBT, Nitro BT
Linear Formula:
C40H30N10O6 · 2Cl
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Application
Technique
Showing 1-3 of 3