![Monoclonal ANTI-FLAG® M2-Alkaline Phosphatase antibody produced in mouse clone M2, purified immunoglobulin, buffered aqueous glycerol solution](/deepweb/assets/sigmaaldrich/product/images/269/856/7a3e2f80-ff58-4436-9ff7-6d18cdb79052/640/7a3e2f80-ff58-4436-9ff7-6d18cdb79052.jpg)
Monoclonal ANTI-FLAG® M2-Alkaline Phosphatase antibody produced in mouse
![Monoclonal ANTI-FLAG® M2-Alkaline Phosphatase antibody produced in mouse clone M2, purified immunoglobulin, buffered aqueous glycerol solution](/deepweb/assets/sigmaaldrich/product/images/269/856/7a3e2f80-ff58-4436-9ff7-6d18cdb79052/640/7a3e2f80-ff58-4436-9ff7-6d18cdb79052.jpg)
Synonym(s):
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, Anti-ddddk, Anti-dykddddk
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Antibody Form
Conjugate
Quality Segment
Species Reactivity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing