
Monoclonal Anti-Cathepsin L antibody produced in mouse

Synonym(s):
Anti-CATL, Anti-CTSL, Anti-MEP
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Antibody Form
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2