
METHYL BESYLATE

Synonym(s):
Methyl benzenesulfonate
Empirical Formula (Hill Notation):
C7H8O3S
CAS Number:
Molecular Weight:
172.20
Products
Formula Weight
Manufacturer
Agency
Format
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing