
Memantine Related Compound B

Synonym(s):
3,5-Dimethyl-1-adamantanol, 3,5-Dimethyl-1-hydroxyadamantane
Empirical Formula (Hill Notation):
C12H20O
CAS Number:
Molecular Weight:
180.29
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2