
Kaempferol 3-rhamnoside

Synonym(s):
Afzelin, 3,4′,5,7-Tetrahydroxyflavone 3-rhamnoside, Afzeloside, Kaempferin, Kaempferol 3-O-alpha-L-rhamnoside, Kaempferol 3-rhamnoside
Empirical Formula (Hill Notation):
C21H20O10
CAS Number:
Molecular Weight:
432.38
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing