
Ibotenic acid

Synonym(s):
α-Amino-3-hydroxy-5-isoxazoleacetic acid, Ibotenic Acid
Empirical Formula (Hill Notation):
C5H6N2O4
CAS Number:
Molecular Weight:
158.11
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing