
Hydrocodone bitartrate salt hemi(pentahydrate)

Empirical Formula (Hill Notation):
C18H21NO3 · C4H6O6 · 2.5H2O
CAS Number:
Molecular Weight:
494.49
Products
Brand
Formula Weight
Agency
Application
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing