
Harmaline solution

Synonym(s):
Harmaline, 1-Methyl-7-methoxy-3,4-dihydro-β-carboline, 3,4-Dihydroharmine, 4,9-Dihydro-7-methoxy-1-methyl-3H-pyrido[3,4-b]indole, Dihydroharmine, Harmalol methyl ether, Harmidine, NSC 407285
Empirical Formula (Hill Notation):
C13H14N2O
CAS Number:
Molecular Weight:
214.26
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Format
Quality Segment
Markush Class
Markush Group
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing