![Glimepiride cis-Isomer (Glimepiride Related Compound A) Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/162/932/905d6557-80e5-43ec-8984-2e23d3ddb547/640/905d6557-80e5-43ec-8984-2e23d3ddb547.jpg)
Glimepiride cis-Isomer (Glimepiride Related Compound A)
![Glimepiride cis-Isomer (Glimepiride Related Compound A) Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/162/932/905d6557-80e5-43ec-8984-2e23d3ddb547/640/905d6557-80e5-43ec-8984-2e23d3ddb547.jpg)
Synonym(s):
3-Ethyl-2,5-dihydro-4-methyl-N-[2-[4-[[[[(cis-4-methylcyclohexyl)amino]carbonyl]amino]sulfonyl]phenyl]ethyl]-2-oxo-1H-pyrrole-1-carboxamide, 1-[[p-[2-(3-Ethyl-4-methyl-2-oxo-3-pyrroline-1-carboxamido)ethyl]phenyl]sulfonyl]-3-(cis-4-methylcyclohexyl)urea, Glimepiride cis-isomer
Empirical Formula (Hill Notation):
C24H34N4O5S
CAS Number:
Molecular Weight:
490.62
Products
Brand
Formula Weight
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing