
Gemcitabine hydrochloride

Synonym(s):
2′-Deoxy-2′,2′-difluorocytidine; dFdC, Gemzar (Lilly), LY-188011, dFdC, dFdCyd
Empirical Formula (Hill Notation):
C9H11F2N3O4 · HCl
CAS Number:
Molecular Weight:
299.66
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-4 of 4
Compare
Product Number
Description
Pricing