
Gabapentin

Synonym(s):
1-(Aminomethyl)-cyclohexaneacetic acid, Neurontin, Gabapentin
Empirical Formula (Hill Notation):
C9H17NO2
CAS Number:
Molecular Weight:
171.24
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing